Assay Details
Target Gene Details
Entrez Gene ID: | 150696 |
Gene Name: | prominin 2 |
Gene Aliases: |
PROML2 |
Location: |
Chr.2:95274448-95291320 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PROM2 | NM_001165977.2 | 7 | 974 | NP_001159449.1 |
| NM_001165978.2 | 7 | 974 | NP_001159450.1 | |
| NM_001321070.1 | 7 | 1035 | NP_001307999.1 | |
| NM_144707.3 | 7 | 974 | NP_653308.2 | |
| XM_006712301.2 | 4 | 304 | XP_006712364.1 | |
| XM_011510672.2 | 7 | 958 | XP_011508974.1 | |
| AF245303.1 | 7 | 969 | AAM10541.1 | |
| AF245304.1 | 7 | 969 | AAM10542.1 | |
| AK091175.1 | 7 | 944 | BAC03599.1 | |
| AK290366.1 | 7 | 929 | ||
| AK313267.1 | 7 | 930 | ||
| AY358377.1 | 7 | 925 | AAQ88743.1 | |
| BC113877.1 | 5 | 795 | AAI13878.1 | |
| BC114525.1 | 7 | 949 | AAI14526.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528011 | Chr.2:95265470 - 95391263 on Build GRCh38 | Loss |
|
| nsv1160825 | Chr.2:95050220 - 95445574 on Build GRCh38 | Duplication |
|
| nsv998014 | Chr.2:95247530 - 95630882 on Build GRCh38 | Gain |
|
| nsv953478 | Chr.2:95266753 - 95326752 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73625087] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map