Assay Details
Target Gene Details
Entrez Gene ID: | 57053 |
Gene Name: | cholinergic receptor nicotinic alpha 10 subunit |
Gene Aliases: |
- |
Location: |
Chr.11:3665587-3673208 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CHRNA10 | NM_001303034.1 | 5 | 1124 | NP_001289963.1 |
NM_001303035.1 | 5 | 1057 | NP_001289964.1 | |
NM_020402.3 | 5 | 1062 | NP_065135.2 | |
XM_011520234.1 | 4 | 4258 | XP_011518536.1 | |
AF199235.2 | 5 | 1062 | AAG00795.2 | |
AJ278118.1 | 5 | 1032 | CAC20435.1 | |
AJ295237.1 | 5 | 1032 | CAC16144.1 | |
BC136250.1 | 5 | 1040 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2759796 | Chr.11:3215282 - 3780199 on Build GRCh38 | Gain+Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv526441 | Chr.11:3604947 - 3735456 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv428247 | Chr.11:3595654 - 3780199 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() |
nsv832051 | Chr.11:3502717 - 3707484 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() |
nsv1046356 | Chr.11:3607607 - 3752985 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
esv3625147 | Chr.11:3607625 - 3732043 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |
nsv482156 | Chr.11:3665587 - 3671384 on Build GRCh38 | Loss |
![]() ![]() |
esv3625148 | Chr.11:3609614 - 3680474 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
