Assay Details
Target Gene Details
Entrez Gene ID: | 3925 |
Gene Name: | stathmin 1 |
Gene Aliases: |
C1orf215, LAP18, Lag, OP18, PP17, PP19, PR22, SMN |
Location: |
Chr.1:25884186-25906877 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STMN1 | NM_001145454.2 | NP_001138926.1 | ||
| NM_005563.3 | 5 | 1263 | NP_005554.1 | |
| NM_203399.1 | 5 | 1239 | NP_981944.1 | |
| NM_203401.1 | 5 | 1451 | NP_981946.1 | |
| AK054594.1 | 6 | 1159 | ||
| AK303692.1 | ||||
| BQ772197.1 | 1 | 280 | ||
| J04991.1 | 5 | 1190 | AAA59980.1 | |
| X53305.1 | 5 | 1168 | CAA37391.1 | |
| Z11566.1 | 5 | 1178 | CAA77660.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834491 | Chr.1:25726411 - 25914117 on Build GRCh38 | Loss |
|
| nsv834502 | Chr.1:25834281 - 25987428 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map