Assay Details
Target Gene Details
Entrez Gene ID: | 23268 |
Gene Name: | dynamin binding protein |
Gene Aliases: |
ARHGEF36, TUBA |
Location: |
Chr.10:99875571-100009953 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DNMBP | NM_001318326.1 | 10 | 2588 | NP_001305255.1 |
| NM_001318327.1 | 10 | 1339 | NP_001305256.1 | |
| NM_015221.3 | 13 | 3585 | NP_056036.1 | |
| XM_006717735.2 | 12 | 3451 | XP_006717798.1 | |
| XM_011539559.2 | 14 | 3656 | XP_011537861.1 | |
| AB023227.1 | 10 | 2675 | BAA76854.1 | |
| AK128576.1 | 10 | 2587 | ||
| AK303479.1 | 5 | 686 | ||
| AL833283.1 | 13 | 3551 | ||
| AY196211.1 | 12 | 3459 | AAP34307.1 | |
| BC041628.1 | 10 | 1328 | AAH41628.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831960 | Chr.10:99833844 - 99995448 on Build GRCh38 | Gain |
|
| nsv1046030 | Chr.10:99831525 - 99887655 on Build GRCh38 | Loss |
|
| nsv552045 | Chr.10:99862766 - 100074335 on Build GRCh38 | Gain |
|
| nsv1046825 | Chr.10:99861014 - 99912354 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map