Assay Details
Target Gene Details
Entrez Gene ID: | 140683 |
Gene Name: | BPI fold containing family A member 2 |
Gene Aliases: |
C20orf70, PSP, SPLUNC2, bA49G10.1 |
Location: |
Chr.20:33161848-33181417 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BPIFA2 | NM_001319164.1 | NP_001306093.1 | ||
| NM_080574.3 | NP_542141.1 | |||
| AK291854.1 | ||||
| AY359055.1 | AAQ89414.1 | |||
| BC065726.1 | AAH65726.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056411 | Chr.20:32469234 - 33355029 on Build GRCh38 | Loss |
|
| esv2751912 | Chr.20:33179619 - 33248657 on Build GRCh38 | Loss |
|
| esv3645678 | Chr.20:33177873 - 33190004 on Build GRCh38 | Loss |
|
| nsv1060241 | Chr.20:33172902 - 33244962 on Build GRCh38 | Loss |
|
| nsv458964 | Chr.20:33152998 - 33229798 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map