Assay Details
Target Gene Details
Entrez Gene ID: | 9554 |
Gene Name: | SEC22 homolog B, vesicle trafficking protein (gene/pseudogene) |
Gene Aliases: |
ERS-24, SEC22L1 |
Location: |
Chr.1:120152520-120176534 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEC22B | NM_004892.5 | NP_004883.3 | ||
| XM_017002856.1 | XP_016858345.1 | |||
| AK289875.1 | ||||
| BC001364.2 | AAH01364.1 | |||
| CD619408.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv957355 | Chr.1:120175848 - 120179977 on Build GRCh38 | Deletion |
|
| nsv161167 | Chr.1:120175846 - 120179977 on Build GRCh38 | Deletion |
|
| nsv1078169 | Chr.1:120175839 - 120179943 on Build GRCh38 | Deletion |
|
| esv2849791 | Chr.1:120175845 - 120179977 on Build GRCh38 | Deletion |
|
| dgv7e180 | Chr.1:120175834 - 120179977 on Build GRCh38 | Deletion |
|
| nsv524681 | Chr.1:118890895 - 121570001 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map