Assay Details
Target Gene Details
Entrez Gene ID: | 23072 |
Gene Name: | HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1 |
Gene Aliases: |
NEDL1 |
Location: |
Chr.7:43112580-43566001 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HECW1 | NM_001287059.1 | NP_001273988.1 | ||
| NM_015052.4 | NP_055867.3 | |||
| XM_005249665.3 | XP_005249722.1 | |||
| XM_006715670.3 | XP_006715733.1 | |||
| XM_006715671.3 | XP_006715734.1 | |||
| XM_006715673.3 | XP_006715736.1 | |||
| XM_017011883.1 | XP_016867372.1 | |||
| XM_017011885.1 | XP_016867374.1 | |||
| XM_017011886.1 | XP_016867375.1 | |||
| XM_017011890.1 | XP_016867379.1 | |||
| AB002320.2 | ||||
| AK294918.1 | ||||
| BC151227.1 |
Target Gene Details
Entrez Gene ID: | 105375254 |
Gene Name: | uncharacterized LOC105375254 |
Gene Aliases: |
- |
Location: |
Chr.7:43025325-43114052 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105375254 | XM_017012919.1 | XP_016868408.1 |
More Information
Set Membership: |
|

Genomic Map