Assay Details
Target Gene Details
Entrez Gene ID: | 122809 |
Gene Name: | suppressor of cytokine signaling 4 |
Gene Aliases: |
SOCS7 |
Location: |
Chr.14:55027126-55049489 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOCS4 | NM_080867.2 | 2 | 1235 | NP_543143.1 |
| NM_199421.1 | 3 | 1364 | NP_955453.1 | |
| XM_011536425.1 | 3 | 1418 | XP_011534727.1 | |
| XM_011536426.1 | 3 | 1333 | XP_011534728.1 | |
| AF424815.1 | 1 | 1015 | AAL60517.1 | |
| AK124083.1 | 2 | 1088 | ||
| AK127299.1 | 1 | 113 | ||
| BC060790.1 | 3 | 1242 | AAH60790.1 | |
| BX161418.1 | 2 | 1131 | CAD61893.1 | |
| BX648734.1 | 2 | 1021 | ||
| CR933659.1 | 2 | 1125 | CAI45958.1 |
More Information
Set Membership: |
|

Genomic Map