Assay Details
Target Gene Details
Entrez Gene ID: | 23033 |
Gene Name: | dopey family member 1 |
Gene Aliases: |
DOP1, KIAA1117, dJ202D23.2 |
Location: |
Chr.6:83067650-83171351 on Build GRCh38 |
Assay Gene Location: | Within Exon 41 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DOPEY1 | NM_001199942.1 | 40 | 7754 | NP_001186871.1 |
| NM_015018.3 | 39 | 7721 | NP_055833.2 | |
| XM_011535619.2 | 39 | 7746 | XP_011533921.1 | |
| XM_017010559.1 | 40 | 7776 | XP_016866048.1 | |
| XM_017010560.1 | 39 | 7704 | XP_016866049.1 | |
| XM_017010561.1 | 40 | 7728 | XP_016866050.1 | |
| XM_017010562.1 | 39 | 7677 | XP_016866051.1 | |
| XM_017010563.1 | 39 | 7695 | XP_016866052.1 | |
| XM_017010564.1 | 39 | 7695 | XP_016866053.1 | |
| XM_017010565.1 | 39 | 7689 | XP_016866054.1 | |
| XM_017010566.1 | 39 | 7689 | XP_016866055.1 | |
| XM_017010567.1 | 38 | 7617 | XP_016866056.1 | |
| XM_017010568.1 | 38 | 7686 | XP_016866057.1 | |
| XM_017010569.1 | 37 | 7603 | XP_016866058.1 | |
| XM_017010570.1 | 39 | 7677 | XP_016866059.1 | |
| XM_017010571.1 | 39 | 7671 | XP_016866060.1 | |
| XM_017010572.1 | 39 | 7626 | XP_016866061.1 | |
| AB029040.2 | 22 | 4742 | BAA83069.2 | |
| AK027030.1 | 19 | 3488 | BAB15631.1 | |
| AK092929.1 | 8 | 1382 | ||
| AL050080.1 | 11 | 1450 | CAB43259.1 | |
| BC048342.1 | 13 | 1734 | AAH48342.1 | |
| BC144544.1 | 39 | 7592 | ||
| BC150619.1 | 39 | 7592 | ||
| BX648667.1 | 39 | 7719 |
Target Gene Details
Entrez Gene ID: | 5238 |
Gene Name: | phosphoglucomutase 3 |
Gene Aliases: |
AGM1, IMD23, PAGM, PGM 3 |
Location: |
Chr.6:83150728-83193936 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PGM3 | NM_001199917.1 | 14 | 2886 | NP_001186846.1 |
| NM_001199918.1 | 12 | 2548 | NP_001186847.1 | |
| NM_001199919.1 | NP_001186848.1 | |||
| NM_015599.2 | 13 | 2750 | NP_056414.1 | |
| XM_017010935.1 | XP_016866424.1 | |||
| XM_017010936.1 | XP_016866425.1 | |||
| XM_017010937.1 | XP_016866426.1 | |||
| XM_017010938.1 | XP_016866427.1 | |||
| AK023709.1 | BAB14652.1 | |||
| AK225363.1 | ||||
| AL117443.1 | 4 | 1461 | CAB55928.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1026095 | Chr.6:83040907 - 83209453 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map