Assay Details
Target Gene Details
Entrez Gene ID: | 7005 |
Gene Name: | TEA domain transcription factor 3 |
Gene Aliases: |
DTEF-1, ETFR-1, TEAD-3, TEAD5, TEF-5, TEF5 |
Location: |
Chr.6:35473597-35497084 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TEAD3 | NM_003214.3 | 13 | 2440 | NP_003205.2 |
| AF142482.1 | 13 | 2394 | AAD29052.1 | |
| AK226144.1 | 9 | 3800 | ||
| BC010053.2 | 13 | 1377 | AAH10053.2 | |
| BC027877.1 | 11 | 2178 | AAH27877.1 | |
| BC091488.1 | 13 | 2365 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528351 | Chr.6:35410145 - 35498009 on Build GRCh38 | Loss |
|
| nsv602910 | Chr.6:35411001 - 35489619 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map