Assay Details
Target Gene Details
Entrez Gene ID: | 9128 |
Gene Name: | pre-mRNA processing factor 4 |
Gene Aliases: |
HPRP4, HPRP4P, PRP4, Prp4p, RP70, SNRNP60 |
Location: |
Chr.9:113275634-113292905 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRPF4 | NM_001244926.1 | NP_001231855.1 | ||
| NM_001322266.1 | NP_001309195.1 | |||
| NM_001322267.1 | NP_001309196.1 | |||
| NM_004697.4 | NP_004688.2 | |||
| NR_136265.1 | ||||
| NR_136266.1 | ||||
| AB209797.1 | BAD93034.1 | |||
| AF016369.1 | AAC51925.1 | |||
| AK225056.1 | ||||
| AK313131.1 | ||||
| AY572489.1 | AAS78572.1 | |||
| BC001588.2 | AAH01588.1 | |||
| BC007424.2 | AAH07424.1 | |||
| CR457105.1 | CAG33386.1 | |||
| DA815191.1 | ||||
| U82756.1 | AAC02261.1 |
More Information
Set Membership: |
|

Genomic Map