Assay Details
Target Gene Details
Entrez Gene ID: | 81624 |
Gene Name: | diaphanous related formin 3 |
Gene Aliases: |
AN, AUNA1, DIA2, DRF3, NSDAN, diap3, mDia2 |
Location: |
Chr.13:59665583-60163985 on Build GRCh38 |
Assay Gene Location: | Within Exon 33 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIAPH3 | NM_001042517.1 | 28 | 3617 | NP_001035982.1 |
| NM_001258366.1 | 27 | 3584 | NP_001245295.1 | |
| NM_001258367.1 | 26 | 3479 | NP_001245296.1 | |
| NM_001258368.1 | 26 | 3407 | NP_001245297.1 | |
| XM_006719876.1 | 22 | 2768 | XP_006719939.1 | |
| AB244756.1 | 27 | 3365 | ||
| AB244757.1 | 26 | 3260 | ||
| AB244758.1 | 26 | 3188 | ||
| AY750055.1 | 28 | 3490 | AAW73254.1 | |
| BC041395.1 | 2 | 186 | AAH41395.1 | |
| BG203073.1 | 4 | 421 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3892363 | Chr.13:58247917 - 59978884 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map