Assay Details
Target Gene Details
Entrez Gene ID: | 84735 |
Gene Name: | carnosine dipeptidase 1 |
Gene Aliases: |
CN1, CPGL2, HsT2308 |
Location: |
Chr.18:74534457-74585025 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNDP1 | NM_032649.5 | 6 | 859 | NP_116038.4 |
| AJ417564.1 | 6 | 692 | CAD10388.1 | |
| AK098369.1 | 7 | 638 | ||
| AK289916.1 | 6 | 714 | ||
| AK303708.1 | 6 | 668 | ||
| AK308716.1 | 6 | 812 | ||
| AK314572.1 | 6 | 750 | ||
| AY358756.1 | 6 | 848 | AAQ89116.1 | |
| BC110295.1 | 6 | 698 | AAI10296.1 | |
| BC113512.1 | 6 | 687 | ||
| BC117122.1 | 6 | 690 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057441 | Chr.18:74344906 - 74669985 on Build GRCh38 | Gain |
|
| esv3643117 | Chr.18:74454714 - 74614537 on Build GRCh38 | Loss |
|
| nsv577656 | Chr.18:74534683 - 75005167 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map