Assay Details
Target Gene Details
Entrez Gene ID: | 5110 |
Gene Name: | protein-L-isoaspartate (D-aspartate) O-methyltransferase |
Gene Aliases: |
PIMT |
Location: |
Chr.6:149749695-149811421 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCMT1 | NM_001252049.1 | NP_001238978.1 | ||
| NM_001252050.1 | NP_001238979.1 | |||
| NM_001252051.1 | NP_001238980.1 | |||
| NM_001252052.1 | NP_001238981.1 | |||
| NM_001252053.1 | NP_001238982.1 | |||
| NM_005389.2 | NP_005380.2 | |||
| AK098739.1 | ||||
| AK129778.1 | ||||
| AK289724.1 | ||||
| AK304532.1 | ||||
| AL539512.3 | ||||
| BC007501.2 | AAH07501.1 | |||
| BC008748.2 | AAH08748.1 | |||
| BE262147.1 | ||||
| BX438393.2 | ||||
| CX786396.1 | ||||
| D13892.1 | BAA02991.1 | |||
| D25545.1 | BAA05028.1 | |||
| D25546.1 | BAA05029.1 | |||
| D25547.1 | BAA05030.1 | |||
| DB478200.1 | ||||
| DC339361.1 | ||||
| DC356391.1 | ||||
| DC421281.1 | ||||
| M93008.1 | AAA90934.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2732861 | Chr.6:149681836 - 149764933 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map