Assay Details
Target Gene Details
Entrez Gene ID: | 22933 |
Gene Name: | sirtuin 2 |
Gene Aliases: |
SIR2, SIR2L, SIR2L2 |
Location: |
Chr.19:38878555-38899862 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIRT2 | NM_001193286.1 | NP_001180215.1 | ||
| NM_012237.3 | NP_036369.2 | |||
| NM_030593.2 | NP_085096.1 | |||
| NR_034146.1 | ||||
| XM_006723111.1 | XP_006723174.1 | |||
| XM_011526654.1 | XP_011524956.1 | |||
| XM_011526655.1 | XP_011524957.1 | |||
| XM_017026500.1 | XP_016881989.1 | |||
| AB009274.1 | ||||
| AF083107.2 | AAD40850.2 | |||
| AF095714.1 | AAD45971.1 | |||
| AF131800.1 | AAD20046.1 | |||
| AF160214.1 | AAF67015.1 | |||
| AJ505014.1 | CAD43717.1 | |||
| AK025876.1 | ||||
| AK054642.1 | ||||
| AK131390.1 | 5 | 3643 | ||
| AK225824.1 | ||||
| AK290716.1 | ||||
| AK314492.1 | ||||
| AY030277.1 | AAK51133.1 | |||
| BC003012.1 | AAH03012.1 | |||
| BC003547.1 | AAH03547.1 | |||
| KF032391.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160616 | Chr.19:38839331 - 38903590 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map