Assay Details
Target Gene Details
Entrez Gene ID: | 22859 |
Gene Name: | adhesion G protein-coupled receptor L1 |
Gene Aliases: |
CIRL1, CL1, LEC2, LPHN1 |
Location: |
Chr.19:14147737-14206245 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRL1 | NM_001008701.2 | 24 | 6979 | NP_001008701.1 |
| NM_014921.4 | 23 | 6964 | NP_055736.2 | |
| XM_005259818.3 | 23 | 7092 | XP_005259875.1 | |
| XM_011527796.2 | 24 | 7128 | XP_011526098.1 | |
| XM_011527798.2 | 23 | 7113 | XP_011526100.1 | |
| XM_011527801.2 | 22 | 6568 | XP_011526103.1 | |
| XM_017026475.1 | 24 | 7131 | XP_016881964.1 | |
| XM_017026476.1 | 25 | 7241 | XP_016881965.1 | |
| XM_017026477.1 | 24 | 7110 | XP_016881966.1 | |
| XM_017026478.1 | 24 | 7059 | XP_016881967.1 | |
| XM_017026479.1 | 23 | 7041 | XP_016881968.1 | |
| XM_017026480.1 | 21 | 6546 | XP_016881969.1 | |
| AK022001.1 | 1 | 1347 | BAB13950.1 |
Target Gene Details
Entrez Gene ID: | 100507373 |
Gene Name: | uncharacterized LOC100507373 |
Gene Aliases: |
- |
Location: |
Chr.19:14137152-14171264 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100507373 | NR_045214.1 | |||
| BX537706.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953980 | Chr.19:14082589 - 14210288 on Build GRCh38 | Deletion |
|
| dgv49n68 | Chr.19:14066502 - 14274131 on Build GRCh38 | Loss |
|
| nsv517436 | Chr.19:14135617 - 14187630 on Build GRCh38 | Loss |
|
| nsv578642 | Chr.19:14028192 - 14187630 on Build GRCh38 | Gain |
|
| nsv578663 | Chr.19:14111697 - 14226588 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs73509752] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map