Assay Details
Target Gene Details
Entrez Gene ID: | 2585 |
Gene Name: | galactokinase 2 |
Gene Aliases: |
GK2 |
Location: |
Chr.15:49155757-49367883 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GALK2 | NM_001001556.2 | 1 | 174 | NP_001001556.1 |
| XM_005254280.3 | 1 | 156 | XP_005254337.1 | |
| XM_005254284.3 | 1 | 156 | XP_005254341.1 | |
| XM_017022063.1 | 1 | 156 | XP_016877552.1 | |
| XM_017022065.1 | 1 | 176 | XP_016877554.1 | |
| AF461816.1 | 1 | 154 | AAP97708.1 | |
| AL832745.1 | 1 | 155 | ||
| DC390737.1 | 1 | 277 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv827332 | Chr.15:49155529 - 49155979 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map