Assay Details
Target Gene Details
Entrez Gene ID: | 2255 |
Gene Name: | fibroblast growth factor 10 |
Gene Aliases: |
- |
Location: |
Chr.5:44301655-44389780 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGF10 | NM_004465.1 | NP_004456.1 | ||
| XM_005248264.3 | 2 | 995 | XP_005248321.1 | |
| AB002097.1 | BAA22331.1 | |||
| AF411527.1 | AAL05875.1 | |||
| AK315314.1 | 1 | 225 | ||
| BC069561.1 | AAH69561.1 | |||
| BC105021.1 | 1 | 70 | AAI05022.1 | |
| BC105023.1 | 1 | 67 | AAI05024.1 | |
| CR541665.1 | CAG46466.1 | |||
| CR541688.1 | CAG46489.1 | |||
| GQ351295.1 | ||||
| U67918.1 | AAB61991.1 |
Target Gene Details
Entrez Gene ID: | 101927075 |
Gene Name: | FGF10 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.5:44388732-44413989 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGF10-AS1 | NR_108034.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830277 | Chr.5:44383300 - 44566779 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map