Assay Details
Target Gene Details
Entrez Gene ID: | 10869 |
Gene Name: | ubiquitin specific peptidase 19 |
Gene Aliases: |
ZMYND9 |
Location: |
Chr.3:49108046-49120938 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834686 | Chr.3:49068614 - 49233123 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map