Assay Details
Target Gene Details
Entrez Gene ID: | 23310 |
Gene Name: | non-SMC condensin II complex subunit D3 |
Gene Aliases: |
CAP-D3, CAPD3, hCAP-D3, hHCP-6, hcp-6 |
Location: |
Chr.11:134152442-134225443 on Build GRCh38 |
Assay Gene Location: | Within Exon 35 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NCAPD3 | NM_015261.2 | 35 | 5125 | NP_056076.1 |
| XM_011542719.2 | 34 | 4386 | XP_011541021.1 | |
| AF070553.1 | 9 | 1038 | AAC28639.1 | |
| AK025549.1 | 9 | 1155 | ||
| AK025667.1 | 19 | 2416 | ||
| AK095267.1 | 5 | 2268 | ||
| AK124878.1 | 35 | 5051 | ||
| BC011408.1 | 13 | 1700 | ||
| BC098398.1 | 35 | 4584 | AAH98398.1 | |
| D29954.1 | 35 | 4547 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521581 | Chr.11:134126584 - 134772738 on Build GRCh38 | Gain |
|
| esv3892114 | Chr.11:134054794 - 134328278 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map