Assay Details
Target Gene Details
Entrez Gene ID: | 57521 |
Gene Name: | regulatory associated protein of MTOR complex 1 |
Gene Aliases: |
KOG1, Mip1 |
Location: |
Chr.17:80544825-80966373 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPTOR | NM_001163034.1 | NP_001156506.1 | ||
| NM_020761.2 | NP_065812.1 | |||
| AI821182.1 | ||||
| AK295964.1 | ||||
| AK296321.1 | ||||
| BC064515.1 | AAH64515.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1056989 | Chr.17:80534340 - 80557095 on Build GRCh38 | Gain |
|
| esv24254 | Chr.17:80396528 - 80656857 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map