Assay Details
Target Gene Details
Entrez Gene ID: | 9869 |
Gene Name: | SET domain bifurcated 1 |
Gene Aliases: |
ESET, H3-K9-HMTase4, KG1T, KMT1E, TDRD21 |
Location: |
Chr.1:150926246-150964744 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SETDB1 | NM_001145415.1 | 13 | 1874 | NP_001138887.1 |
| NM_012432.3 | 13 | 1874 | NP_036564.3 | |
| XM_005245641.3 | 13 | 1877 | XP_005245698.1 | |
| XM_017002953.1 | 13 | 1801 | XP_016858442.1 | |
| XM_017002954.1 | 13 | 1877 | XP_016858443.1 | |
| AK308335.1 | 13 | 1821 | ||
| BC028671.2 | 13 | 1801 | AAH28671.1 | |
| D31891.1 | 13 | 1770 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1008496 | Chr.1:150873351 - 150998180 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map