Assay Details
Target Gene Details
Entrez Gene ID: | 51166 |
Gene Name: | aminoadipate aminotransferase |
Gene Aliases: |
KAT2, KATII, KYAT2 |
Location: |
Chr.4:170060222-170091699 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AADAT | NM_001286682.1 | NP_001273611.1 | ||
| NM_001286683.1 | NP_001273612.1 | |||
| NM_016228.3 | NP_057312.1 | |||
| NM_182662.1 | NP_872603.1 | |||
| XM_006714231.2 | XP_006714294.1 | |||
| XM_011532020.2 | XP_011530322.1 | |||
| AF481738.1 | AAM09683.1 | |||
| AI190763.1 | ||||
| AK055952.1 | ||||
| AK314729.1 | ||||
| BC031068.1 | AAH31068.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4601 | Chr.4:170048721 - 170093413 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map