Assay Details
Target Gene Details
Entrez Gene ID: | 10961 |
Gene Name: | endoplasmic reticulum protein 29 |
Gene Aliases: |
C12orf8, ERp28, ERp31, HEL-S-107, PDI-DB, PDIA9 |
Location: |
Chr.12:112013340-112023220 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERP29 | NM_001034025.1 | NP_001029197.1 | ||
| NM_006817.3 | NP_006808.1 | |||
| XM_017018720.1 | XP_016874209.1 | |||
| AA412124.1 | ||||
| BC101493.1 | AAI01494.1 | |||
| BC101495.1 | AAI01496.1 | |||
| CR541667.1 | CAG46468.1 | |||
| FJ224317.1 | ||||
| X94910.1 | CAA64397.1 |
More Information
Set Membership: |
|

Genomic Map