Assay Details
Target Gene Details
Entrez Gene ID: | 2585 |
Gene Name: | galactokinase 2 |
Gene Aliases: |
GK2 |
Location: |
Chr.15:49155757-49367883 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636423 | Chr.15:49208792 - 49259459 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map