Assay Details
Target Gene Details
Entrez Gene ID: | 28964 |
Gene Name: | GIT ArfGAP 1 |
Gene Aliases: |
- |
Location: |
Chr.17:29573469-29589592 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GIT1 | NM_001085454.1 | NP_001078923.1 | ||
| NM_014030.3 | NP_054749.2 | |||
| XM_011524684.1 | XP_011522986.1 | |||
| XM_011524685.1 | XP_011522987.1 | |||
| AB209537.1 | BAD92774.1 | |||
| AF124490.1 | AAD28046.1 | |||
| AK000863.1 | ||||
| AK024826.1 | ||||
| AK294785.1 | ||||
| AK297443.1 | ||||
| AK299615.1 | ||||
| AK299932.1 | ||||
| BC006227.2 | AAH06227.2 | |||
| BC032098.1 | AAH32098.1 | |||
| BC048196.1 | AAH48196.1 | |||
| BX537553.1 | CAD97779.1 | |||
| BX537554.1 | CAD97780.1 | |||
| BX648097.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521557 | Chr.17:29562968 - 29595257 on Build GRCh38 | Loss |
|
| nsv833409 | Chr.17:29465646 - 29655807 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map