Assay Details
Target Gene Details
Entrez Gene ID: | 54802 |
Gene Name: | tRNA isopentenyltransferase 1 |
Gene Aliases: |
GRO1, IPPT, IPT, IPTase, MOD5, hGRO1 |
Location: |
Chr.1:39841031-39883511 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRIT1 | NM_001312691.1 | 10 | 1940 | NP_001299620.1 |
| NM_001312692.1 | 9 | 1772 | NP_001299621.1 | |
| NM_017646.5 | 11 | 2018 | NP_060116.2 | |
| NR_132401.1 | 11 | 2014 | ||
| NR_132402.1 | 10 | 1872 | ||
| NR_132403.1 | 10 | 1868 | ||
| NR_132404.1 | 10 | 1868 | ||
| NR_132405.1 | 10 | 1864 | ||
| NR_132406.1 | 9 | 1755 | ||
| NR_132407.1 | 8 | 1632 | ||
| NR_132408.1 | 8 | 1628 | ||
| NR_132409.1 | 7 | 1489 | ||
| NR_132410.1 | 7 | 1515 | ||
| NR_132412.1 | 6 | 1376 | ||
| NR_132413.1 | 4 | 1186 | ||
| NR_132414.1 | 2 | 958 | ||
| NR_132415.1 | 11 | 2105 | ||
| XM_005270954.2 | 11 | 2254 | XP_005271011.1 | |
| XM_006710706.1 | 11 | 2086 | XP_006710769.1 | |
| AF074918.1 | 11 | 2099 | AAG31324.1 | |
| AK000068.1 | 9 | 1635 | BAA90923.1 | |
| AK074222.1 | 9 | 1635 | ||
| AK223478.1 | 11 | 2011 | BAD97198.1 | |
| AY303390.1 | 11 | 2012 | AAP60111.1 | |
| BC010741.1 | 9 | 1734 | AAH10741.2 | |
| BC107569.1 | 11 | 2017 | AAI07570.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2758933 | Chr.1:39759471 - 40194760 on Build GRCh38 | Loss |
|
| dgv153n100 | Chr.1:39715819 - 39848248 on Build GRCh38 | Gain |
|
| esv3585792 | Chr.1:39811141 - 39845795 on Build GRCh38 | Loss |
|
| nsv546063 | Chr.1:39719061 - 39841226 on Build GRCh38 | Gain |
|
| esv2666580 | Chr.1:39809257 - 39898016 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map