Assay Details
Target Gene Details
Entrez Gene ID: | 285175 |
Gene Name: | unc-80 homolog, NALCN activator |
Gene Aliases: |
C2orf21, UNC-80 |
Location: |
Chr.2:209771925-209999300 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UNC80 | NM_032504.1 | 3 | 272 | NP_115893.1 |
| NM_182587.3 | 3 | 272 | NP_872393.3 | |
| XM_005246476.1 | 3 | 272 | XP_005246533.1 | |
| XM_011511004.1 | 3 | 272 | XP_011509306.1 | |
| XM_011511005.1 | 3 | 272 | XP_011509307.1 | |
| XM_011511006.1 | 3 | 272 | XP_011509308.1 | |
| XM_011511007.1 | 3 | 272 | XP_011509309.1 | |
| XM_011511008.1 | 3 | 272 | XP_011509310.1 | |
| XM_011511010.2 | 3 | 272 | XP_011509312.1 | |
| XM_011511012.1 | 3 | 272 | XP_011509314.1 | |
| XM_017003884.1 | 3 | 272 | XP_016859373.1 | |
| XM_017003885.1 | 3 | 272 | XP_016859374.1 | |
| XM_017003886.1 | 3 | 272 | XP_016859375.1 | |
| XM_017003887.1 | 3 | 272 | XP_016859376.1 | |
| XM_017003888.1 | 3 | 272 | XP_016859377.1 | |
| XM_017003889.1 | 3 | 272 | XP_016859378.1 | |
| XM_017003890.1 | 3 | 272 | XP_016859379.1 | |
| XM_017003891.1 | 2 | 291 | XP_016859380.1 | |
| XM_017003893.1 | 3 | 272 | XP_016859382.1 | |
| AK090815.1 | 4 | 272 | BAC03521.1 | |
| BC136690.1 | 3 | 243 | ||
| BC136693.1 | 3 | 243 | ||
| DA147023.1 | 4 | 272 |
More Information
Set Membership: |
|

Genomic Map