Assay Details
Target Gene Details
Entrez Gene ID: | 1368 |
Gene Name: | carboxypeptidase M |
Gene Aliases: |
- |
Location: |
Chr.12:68842200-68963240 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CPM | NM_001005502.2 | 9 | 4584 | NP_001005502.1 |
| NM_001874.4 | 9 | 4616 | NP_001865.1 | |
| NM_198320.3 | 9 | 4602 | NP_938079.1 | |
| BQ181238.1 | ||||
| BQ706414.1 | 1 | 645 |
More Information
Set Membership: |
|

Genomic Map