Assay Details
Target Gene Details
Entrez Gene ID: | 5159 |
Gene Name: | platelet derived growth factor receptor beta |
Gene Aliases: |
CD140B, IBGC4, IMF1, JTK12, KOGS, PDGFR, PDGFR-1, PDGFR1, PENTT |
Location: |
Chr.5:150113839-150155884 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDGFRB | NM_002609.3 | 23 | 5493 | NP_002600.1 |
| XM_005268464.2 | 22 | 5472 | XP_005268521.1 | |
| XM_011537658.1 | 23 | 5068 | XP_011535960.1 | |
| XM_011537659.1 | 24 | 5973 | XP_011535961.1 | |
| AI346188.1 | 1 | 226 | ||
| BC032224.1 | 23 | 5499 | AAH32224.1 | |
| J03278.1 | 23 | 5380 | AAA60049.1 | |
| M21616.1 | 23 | 5210 | AAA36427.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830518 | Chr.5:149940811 - 150117140 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map