Assay Details
Target Gene Details
Entrez Gene ID: | 57214 |
Gene Name: | cell migration inducing hyaluronan binding protein |
Gene Aliases: |
CCSP1, HYBID, KIAA1199, TMEM2L |
Location: |
Chr.15:80779343-80951662 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CEMIP | NM_001293298.1 | 30 | 6278 | NP_001280227.1 |
NM_001293304.1 | 30 | 6218 | NP_001280233.1 | |
NM_018689.2 | 29 | 6119 | NP_061159.1 | |
AB033025.1 | 21 | 4787 | BAA86513.1 | |
AB103330.1 | 29 | 6091 | BAD02451.1 | |
AK000465.1 | 19 | 3563 | ||
AK026540.1 | 14 | 3668 | ||
AK027058.1 | 1 | 1782 | ||
AL049389.1 | 1 | 1317 | ||
AL109718.1 | 1 | 691 | ||
AL359061.1 | 17 | 4162 | CAB94391.1 | |
AY007811.1 | 10 | 3250 | AAG41059.1 | |
AY581148.1 | 30 | 6250 | AAU06219.1 | |
AY581149.1 | 30 | 6190 | AAU06220.1 | |
AY585237.1 | 29 | 6091 | AAU06221.1 |
Target Gene Details
Entrez Gene ID: | 23184 |
Gene Name: | mesoderm development candidate 2 |
Gene Aliases: |
BOCA, MESD |
Location: |
Chr.15:80947326-80989878 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MESDC2 | NR_126327.1 | |||
NR_126328.1 | ||||
AK301416.1 | ||||
BC009210.2 | AAH09210.1 | |||
BC012746.2 | AAH12746.1 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
