Assay Details
Target Gene Details
Entrez Gene ID: | 80072 |
Gene Name: | HEXA antisense RNA 1 |
Gene Aliases: |
C15orf34 |
Location: |
Chr.15:72376113-72378788 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HEXA-AS1 | NR_027262.1 | 1 | 855 | |
| AK023377.1 | 1 | 603 | BAB14550.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521971 | Chr.15:72374083 - 72386397 on Build GRCh38 | Gain |
|

Genomic Map