Assay Details
Target Gene Details
Entrez Gene ID: | 4204 |
Gene Name: | methyl-CpG binding protein 2 |
Gene Aliases: |
AUTSX3, MRX16, MRX79, MRXS13, MRXSL, PPMX, RS, RTS, RTT |
Location: |
Chr.X:154021800-154097731 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MECP2 | NM_004992.3 | 4 | 6800 | NP_004983.1 |
| XM_005274682.4 | 5 | 6874 | XP_005274739.1 | |
| XM_006724819.3 | 4 | 6551 | XP_006724882.1 | |
| XM_011531166.2 | 3 | 6687 | XP_011529468.1 | |
| XM_017029533.1 | 7 | 7300 | XP_016885022.1 | |
| AB209464.1 | 4 | 5439 | BAD92701.1 | |
| AF158180.1 | 4 | 6741 | AAF33023.1 | |
| AJ132917.1 | 3 | 6649 | CAB46446.1 | |
| BC031833.1 | 1 | 2133 | AAH31833.1 | |
| GQ203293.1 | 4 | 6800 | ||
| GQ203294.1 | 4 | 6800 | ||
| GQ203295.1 | 4 | 6800 | ||
| GQ896382.1 | 4 | 6800 | ||
| GU479943.1 | 4 | 6800 | ||
| HM020402.1 | 4 | 6800 | ||
| HM156732.1 | 4 | 6800 | ||
| HM156733.1 | 4 | 6800 | ||
| HQ141378.1 | 4 | 6800 | ||
| HQ154629.1 | 5 | 6460 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33784 | Chr.X:153962665 - 155699588 on Build GRCh38 | Gain+Loss |
|
| esv33199 | Chr.X:153065734 - 154591978 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map