Assay Details
Target Gene Details
Entrez Gene ID: | 25942 |
Gene Name: | SIN3 transcription regulator family member A |
Gene Aliases: |
- |
Location: |
Chr.15:75369379-75455819 on Build GRCh38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SIN3A | NM_001145357.1 | 21 | 3952 | NP_001138829.1 |
| NM_001145358.1 | 21 | 4127 | NP_001138830.1 | |
| NM_015477.2 | 21 | 4029 | NP_056292.1 | |
| XM_006720465.3 | 21 | 3835 | XP_006720528.1 | |
| XM_006720466.3 | 22 | 4225 | XP_006720529.1 | |
| XM_006720467.3 | 23 | 4043 | XP_006720530.1 | |
| AF418569.1 | 21 | 4029 | AAP97288.1 | |
| AK074800.1 | 7 | 1128 | ||
| AK074903.1 | 9 | 1778 | BAC11280.1 | |
| AL117513.1 | 7 | 1035 | CAB55972.1 | |
| AL832463.1 | 21 | 3953 | ||
| BC018973.1 | 7 | 1202 | AAH18973.1 | |
| BC137098.1 | 21 | 3880 | ||
| BC137099.1 | 21 | 3880 | ||
| BX649162.1 | 21 | 4066 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2621n100 | Chr.15:75101872 - 75382048 on Build GRCh38 | Gain |
|
| esv3636926 | Chr.15:75363580 - 75378339 on Build GRCh38 | Gain |
|
| esv2749894 | Chr.15:75326873 - 75749895 on Build GRCh38 | Deletion |
|
| nsv1615 | Chr.15:75351396 - 75377578 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map