Assay Details
Target Gene Details
Entrez Gene ID: | 5621 |
Gene Name: | prion protein |
Gene Aliases: |
ASCR, AltPrP, CD230, CJD, GSS, KURU, PRIP, PrP, PrP27-30, PrP33-35C, PrPc, p27-30 |
Location: |
Chr.20:4686151-4701588 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRNP | NM_000311.3 | 2 | 428 | NP_000302.1 |
| NM_001080121.1 | 2 | 423 | NP_001073590.1 | |
| NM_001080122.1 | 2 | 419 | NP_001073591.1 | |
| NM_001080123.1 | 2 | 292 | NP_001073592.1 | |
| NM_001271561.1 | 2 | 428 | NP_001258490.1 | |
| NM_183079.2 | 2 | 424 | NP_898902.1 | |
| AB300823.1 | 1 | 66 | ||
| AB300824.1 | 1 | 66 | ||
| AB300825.1 | 1 | 66 | ||
| AB300826.1 | 1 | 66 | ||
| AK090575.1 | 2 | 116 | ||
| AK293312.1 | ||||
| AK295415.1 | ||||
| AK295943.1 | 2 | 116 | ||
| AK312276.1 | 2 | 116 | ||
| AY008282.1 | 2 | 99 | AAG21693.1 | |
| AY569456.1 | 1 | 56 | AAS80162.1 | |
| BC012844.1 | 2 | 103 | AAH12844.1 | |
| BC022532.1 | 2 | 116 | AAH22532.1 | |
| BI669189.1 | 2 | 119 | ||
| BP251427.1 | 2 | 205 | ||
| BT019496.1 | 1 | 56 | AAV38303.1 | |
| CR542039.1 | 1 | 56 | CAG46836.1 | |
| CR542072.1 | 1 | 56 | CAG46869.1 | |
| D00015.1 | 1 | 111 | BAA00011.1 | |
| DA122620.1 | 2 | 368 | ||
| DA297032.1 | 2 | 399 | ||
| DB461478.1 | 2 | 292 | ||
| M13667.1 | 1 | 111 | AAA19664.1 | |
| M13899.1 | 2 | 105 | AAA60182.1 | |
| X82545.1 | CAA57895.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1058830 | Chr.20:4663154 - 4725219 on Build GRCh38 | Gain |
|
| esv3645074 | Chr.20:4205961 - 5037636 on Build GRCh38 | Gain |
|
| nsv585310 | Chr.20:4586645 - 4794039 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map