Assay Details
Target Gene Details
Entrez Gene ID: | 10498 |
Gene Name: | coactivator associated arginine methyltransferase 1 |
Gene Aliases: |
PRMT4 |
Location: |
Chr.19:10871577-10923078 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARM1 | XM_005259708.4 | XP_005259765.1 | ||
| XM_011527638.2 | XP_011525940.1 |
Target Gene Details
Entrez Gene ID: | 78992 |
Gene Name: | Yip1 domain family member 2 |
Gene Aliases: |
FinGER2 |
Location: |
Chr.19:10922185-10929012 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| YIPF2 | NM_001321439.1 | 10 | 1233 | NP_001308368.1 |
| NM_001321440.1 | 9 | 1116 | NP_001308369.1 | |
| NM_024029.4 | 10 | 1504 | NP_076934.1 | |
| XM_011528270.2 | 10 | 1250 | XP_011526572.1 | |
| XM_011528271.2 | 9 | 1094 | XP_011526573.1 | |
| BC000056.2 | 10 | 1152 | AAH00056.1 | |
| BC013014.2 | 10 | 1173 | AAH13014.1 | |
| BE275116.1 | 5 | 596 | ||
| BM554546.1 | ||||
| BX327495.2 | 3 | 371 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833749 | Chr.19:10881255 - 11027578 on Build GRCh38 | Loss |
|
| nsv833748 | Chr.19:10757002 - 10925368 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map