Assay Details
Target Gene Details
Entrez Gene ID: | 105373805 |
Gene Name: | uncharacterized LOC105373805 |
Gene Aliases: |
- |
Location: |
Chr.2:191021526-191032314 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105373805 | NR_136318.1 | |||
| AW873644.1 |
Target Gene Details
Entrez Gene ID: | 6775 |
Gene Name: | signal transducer and activator of transcription 4 |
Gene Aliases: |
SLEB11 |
Location: |
Chr.2:191029576-191172684 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAT4 | NM_001243835.1 | 24 | 2683 | NP_001230764.1 |
| NM_003151.3 | 24 | 2734 | NP_003142.1 | |
| XM_006712719.3 | 24 | 2729 | XP_006712782.1 | |
| XM_011511705.2 | 27 | 2880 | XP_011510007.1 | |
| AK299976.1 | 21 | 2215 | ||
| BC031212.1 | 24 | 2655 | AAH31212.1 | |
| L78440.1 | 24 | 2500 | AAB05605.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv999391 | Chr.2:190884309 - 191057205 on Build GRCh38 | Gain |
|
| nsv834493 | Chr.2:190946647 - 191109514 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map