Assay Details
Target Gene Details
Entrez Gene ID: | 55691 |
Gene Name: | FERM domain containing 4A |
Gene Aliases: |
CCAFCA, FRMD4, bA295P9.4 |
Location: |
Chr.10:13643706-14330952 on Build GRCh38 |
Assay Gene Location: | Within Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FRMD4A | NM_001318336.1 | 21 | 3326 | NP_001305265.1 |
| NM_001318337.1 | 21 | 3566 | NP_001305266.1 | |
| NM_001318338.1 | 11 | 2360 | NP_001305267.1 | |
| NM_018027.4 | 22 | 3266 | NP_060497.3 | |
| XM_005252489.2 | 21 | 3566 | XP_005252546.1 | |
| XM_005252490.2 | 22 | 3362 | XP_005252547.1 | |
| XM_005252495.4 | 8 | 4525 | XP_005252552.1 | |
| XM_006717457.2 | 23 | 3647 | XP_006717520.1 | |
| XM_006717458.3 | 23 | 3647 | XP_006717521.1 | |
| XM_006717459.2 | 22 | 3611 | XP_006717522.1 | |
| XM_006717460.2 | 22 | 3602 | XP_006717523.1 | |
| XM_006717464.1 | 8 | 2026 | XP_006717527.1 | |
| XM_011519539.1 | 23 | 3407 | XP_011517841.1 | |
| XM_011519540.1 | 23 | 3596 | XP_011517842.1 | |
| XM_011519541.1 | 22 | 3590 | XP_011517843.1 | |
| XM_011519543.2 | 23 | 3370 | XP_011517845.1 | |
| XM_017016393.1 | 22 | 3611 | XP_016871882.1 | |
| XM_017016394.1 | 22 | 3334 | XP_016871883.1 | |
| XM_017016395.1 | 21 | 3169 | XP_016871884.1 | |
| XM_017016396.1 | 9 | 2245 | XP_016871885.1 | |
| AB037715.1 | 22 | 3266 | ||
| AK289693.1 | 22 | 3191 | ||
| BC151244.1 | 22 | 3277 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831790 | Chr.10:13536081 - 13716895 on Build GRCh38 | Loss |
|
| esv3891761 | Chr.10:13544627 - 13695972 on Build GRCh38 | Gain |
|
| esv2761575 | Chr.10:13581531 - 13659502 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map