Assay Details
Target Gene Details
Entrez Gene ID: | 57700 |
Gene Name: | family with sequence similarity 160 member B1 |
Gene Aliases: |
KIAA1600, bA106M7.3 |
Location: |
Chr.10:114821744-114899827 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM160B1 | NM_001135051.1 | NP_001128523.1 | ||
| NM_020940.3 | NP_065991.3 | |||
| AB046820.2 | BAB13426.2 | |||
| BC037207.1 | AAH37207.1 | |||
| CR933692.1 | CAI45992.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1038050 | Chr.10:114555350 - 115135954 on Build GRCh38 | Gain |
|

Genomic Map