Assay Details
Target Gene Details
Entrez Gene ID: | 8482 |
Gene Name: | semaphorin 7A (John Milton Hagen blood group) |
Gene Aliases: |
CD108, CDw108, H-SEMA-K1, H-Sema-L, JMH, SEMAK1, SEMAL |
Location: |
Chr.15:74409284-74433959 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEMA7A | NM_001146029.2 | 9 | 1193 | NP_001139501.1 |
| NM_001146030.2 | 10 | 1131 | NP_001139502.1 | |
| NM_003612.4 | 10 | 1235 | NP_003603.1 | |
| AF030698.1 | 10 | 1206 | AAC34261.1 | |
| AF069493.1 | 10 | 1211 | AAC82642.1 | |
| AF071542.1 | 10 | 1194 | AAC80456.1 | |
| AK001895.1 | 4 | 476 | ||
| AK293280.1 | 9 | 1189 | ||
| AK312257.1 | 10 | 1234 | ||
| AM180445.1 | 10 | 1194 | CAJ55398.1 | |
| AM180446.1 | 10 | 1194 | CAJ55399.1 | |
| AM180447.1 | 10 | 1194 | CAJ55400.1 | |
| AM180448.1 | 10 | 1194 | CAJ55401.1 | |
| AM180449.1 | 10 | 1194 | CAJ55402.1 | |
| AM180450.1 | 10 | 1194 | CAJ55403.1 | |
| AM180451.1 | 10 | 1194 | CAJ55404.1 | |
| BC101643.1 | 10 | 1226 | AAI01644.1 | |
| BC101647.1 | 10 | 1226 | AAI01648.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569974 | Chr.15:74408859 - 74434772 on Build GRCh38 | Gain |
|
| nsv1121512 | Chr.15:72670085 - 75273877 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map