Assay Details
Target Gene Details
Entrez Gene ID: | 55893 |
Gene Name: | zinc finger protein 395 |
Gene Aliases: |
HDBP-2, HDBP2, HDRF-2, PBF, PRF-1, PRF1, Si-1-8-14 |
Location: |
Chr.8:28345585-28386460 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF395 | NM_018660.2 | 10 | 2933 | NP_061130.1 |
| AK026076.1 | 1 | 800 | BAB15352.1 | |
| AK312078.1 | 1 | 323 | ||
| AL137588.1 | 1 | 249 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv456n21 | Chr.8:28321496 - 28467894 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map