Assay Details
Target Gene Details
Entrez Gene ID: | 10501 |
Gene Name: | semaphorin 6B |
Gene Aliases: |
SEM-SEMA-Y, SEMA-VIB, SEMAN, semaZ |
Location: |
Chr.19:4542588-4581484 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SEMA6B | NM_032108.3 | NP_115484.2 | ||
| XM_011527639.2 | XP_011525941.1 | |||
| XM_011527640.2 | XP_011525942.1 | |||
| XM_011527641.1 | XP_011525943.1 | |||
| AB022433.1 | BAB20669.1 | |||
| AB209278.1 | BAD92515.1 | |||
| AF216389.1 | AAF87661.1 | |||
| AF293363.1 | AAK16831.1 | |||
| AK300037.1 | ||||
| AY358939.1 | AAQ89298.1 | |||
| BC142617.1 | ||||
| FR839673.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv953950 | Chr.19:4540589 - 4723488 on Build GRCh38 | Deletion |
|
| nsv578444 | Chr.19:4502189 - 4559634 on Build GRCh38 | Gain |
|
| nsv428361 | Chr.19:4510042 - 4689189 on Build GRCh38 | Loss |
|
| nsv2394 | Chr.19:4547422 - 4592423 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map