Assay Details
Target Gene Details
Entrez Gene ID: | 343099 |
Gene Name: | coiled-coil domain containing 18 |
Gene Aliases: |
NY-SAR-41 |
Location: |
Chr.1:93179802-93279037 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC18 | XM_017001164.1 | XP_016856653.1 |
Target Gene Details
Entrez Gene ID: | 50999 |
Gene Name: | transmembrane p24 trafficking protein 5 |
Gene Aliases: |
CGI-100, p24g2, p28 |
Location: |
Chr.1:93149742-93180689 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMED5 | NM_001167830.1 | NP_001161302.1 | ||
| NM_016040.4 | NP_057124.3 | |||
| NR_030761.1 | ||||
| AF151858.1 | AAD34095.1 | |||
| AK125882.1 | ||||
| AK293308.1 | ||||
| AK312787.1 | ||||
| AY359039.1 | AAQ89398.1 | |||
| BC016365.1 | AAH16365.1 | |||
| BC016556.1 | AAH16556.1 | |||
| BC070051.1 | AAH70051.1 | |||
| BX642274.1 | ||||
| CR936808.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830625 | Chr.1:93100821 - 93269689 on Build GRCh38 | Loss |
|
| nsv830614 | Chr.1:93000421 - 93213143 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map