Assay Details
Target Gene Details
Entrez Gene ID: | 144233 |
Gene Name: | BCDIN3 domain containing RNA methyltransferase |
Gene Aliases: |
- |
Location: |
Chr.12:49836043-49843129 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCDIN3D | NM_181708.2 | 2 | 732 | NP_859059.1 |
| AK292194.1 | 2 | 709 | ||
| BC053560.1 | 2 | 695 | AAH53560.1 | |
| CD690663.1 |
Target Gene Details
Entrez Gene ID: | 100286844 |
Gene Name: | BCDIN3D antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.12:49828543-49841154 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCDIN3D-AS1 | NR_027499.1 | 3 | 494 | |
| NR_027500.1 | 3 | 490 | ||
| NR_027501.1 | 3 | 490 | ||
| AI193197.1 | 2 | 159 | ||
| AK055142.1 | 3 | 494 | ||
| CB153643.1 | 3 | 490 | ||
| DA122615.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522972 | Chr.12:49802611 - 50017048 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map