Assay Details
Target Gene Details
Entrez Gene ID: | 121506 |
Gene Name: | endoplasmic reticulum protein 27 |
Gene Aliases: |
C12orf46, PDIA8 |
Location: |
Chr.12:14914027-14938549 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ERP27 | NM_001300784.1 | 4 | 526 | NP_001287713.1 |
| NM_152321.3 | 6 | 719 | NP_689534.1 | |
| AK056677.1 | 6 | 719 | BAB71251.1 | |
| AY358536.1 | 6 | 699 | AAQ88900.1 | |
| BC030218.1 | 6 | 691 | AAH30218.1 | |
| CD389092.1 | 4 | 401 | ||
| CN296050.1 | 4 | 530 |
More Information
Set Membership: |
|

Genomic Map