Assay Details
Target Gene Details
Entrez Gene ID: | 3547 |
Gene Name: | immunoglobulin superfamily member 1 |
Gene Aliases: |
CHTE, IGCD1, IGDC1, INHBP, PGSF2, p120 |
Location: |
Chr.X:131273506-131289457 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGSF1 | NM_001170961.1 | 20 | 4303 | NP_001164432.1 |
| NM_001170962.1 | 19 | 4261 | NP_001164433.1 | |
| NM_001555.4 | 20 | 4288 | NP_001546.2 | |
| XM_011531330.1 | 21 | 4201 | XP_011529632.1 | |
| XM_011531333.1 | 20 | 4054 | XP_011529635.1 | |
| XM_011531334.1 | 18 | 3727 | XP_011529636.1 | |
| AF034198.1 | 19 | 4110 | AAC52057.1 | |
| AK226008.1 | 20 | 4160 | ||
| AK295438.1 | 11 | 2339 | ||
| AK307871.1 | ||||
| AL137369.1 | 4 | 550 | CAB70713.1 | |
| BC063884.1 | 20 | 4124 | AAH63884.1 | |
| Y10523.1 | 19 | 4085 | CAA71535.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522699 | Chr.X:131270380 - 131288648 on Build GRCh38 | Gain |
|
| nsv528179 | Chr.X:131166643 - 131546956 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map