Assay Details
Target Gene Details
Entrez Gene ID: | 2957 |
Gene Name: | general transcription factor IIA subunit 1 |
Gene Aliases: |
TF2A1, TFIIA, TFIIA-42, TFIIAL |
Location: |
Chr.14:81175452-81221231 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTF2A1 | NM_001278940.1 | NP_001265869.1 | ||
| NM_015859.3 | NP_056943.1 | |||
| NM_201595.2 | NP_963889.1 | |||
| BC107155.2 | AAI07156.1 | |||
| BC107156.1 | AAI07157.1 | |||
| D14886.1 | BAA03603.1 | |||
| D14887.1 | BAA03604.1 | |||
| X75383.1 | CAA53151.1 | |||
| X77225.1 | CAA54442.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832841 | Chr.14:81121978 - 81277577 on Build GRCh38 | Gain |
|
| esv3635076 | Chr.14:80715257 - 81672373 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map