Assay Details
Target Gene Details
Entrez Gene ID: | 55250 |
Gene Name: | elongator acetyltransferase complex subunit 2 |
Gene Aliases: |
SHINC-2, STATIP1, StIP |
Location: |
Chr.18:36129874-36177931 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ELP2 | NM_001242875.2 | NP_001229804.1 | ||
| NM_001242876.2 | NP_001229805.1 | |||
| NM_001242877.2 | NP_001229806.1 | |||
| NM_001242878.2 | NP_001229807.1 | |||
| NM_001242879.2 | NP_001229808.1 | |||
| NM_001324465.1 | NP_001311394.1 | |||
| NM_001324466.1 | NP_001311395.1 | |||
| NM_001324467.1 | NP_001311396.1 | |||
| NM_001324468.1 | NP_001311397.1 | |||
| NM_018255.3 | NP_060725.1 | |||
| NR_040110.2 | ||||
| NR_136897.1 | ||||
| NR_136898.1 | ||||
| NR_137173.1 | ||||
| AF332505.1 | AAK97355.1 | |||
| AF403223.1 | AAQ03093.1 | |||
| AK001741.1 | BAA91874.1 | |||
| AK022709.1 | BAB14193.1 | |||
| AK222791.1 | BAD96511.1 | |||
| AK293051.1 | ||||
| AK300201.1 | ||||
| AK302062.1 | ||||
| BC032553.1 | AAH32553.1 | |||
| CR457269.1 | CAG33550.1 | |||
| DC325711.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv576651 | Chr.18:36017567 - 36959615 on Build GRCh38 | Loss |
|
| nsv576652 | Chr.18:36127201 - 36157117 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map