Assay Details
Target Gene Details
Entrez Gene ID: | 27145 |
Gene Name: | filamin A interacting protein 1 |
Gene Aliases: |
FILIP |
Location: |
Chr.6:75294895-75493829 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FILIP1 | NM_001289987.1 | 7 | 4698 | NP_001276916.1 |
| NM_001300866.1 | NP_001287795.1 | |||
| NM_015687.3 | 6 | 4532 | NP_056502.1 | |
| XM_005248713.3 | XP_005248770.1 | |||
| XM_005248715.4 | XP_005248772.1 | |||
| XM_011535756.2 | 3 | 3633 | XP_011534058.1 | |
| AB033101.1 | 4 | 3920 | BAA86589.1 | |
| AB086011.1 | 5 | 4116 | BAC77067.1 | |
| AK001570.1 | 2 | 2463 | BAA91763.1 | |
| AK027705.1 | 2 | 3429 | BAB55310.1 | |
| AL832009.2 | CAD89912.1 | |||
| BX647178.1 | 6 | 4483 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830698 | Chr.6:75259094 - 75405876 on Build GRCh38 | Gain |
|
| nsv603764 | Chr.6:75254035 - 75315887 on Build GRCh38 | Loss |
|

Genomic Map