Assay Details
Target Gene Details
Entrez Gene ID: | 2237 |
Gene Name: | flap structure-specific endonuclease 1 |
Gene Aliases: |
FEN-1, MF1, RAD2 |
Location: |
Chr.11:61792637-61797244 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FEN1 | NM_004111.5 | 1 | 276 | NP_004102.1 |
| AK301743.1 | ||||
| AK312761.1 | ||||
| BC000323.2 | AAH00323.1 | |||
| BP244711.1 | 1 | 256 | ||
| CN290841.1 | 1 | 276 | ||
| X76771.1 | 1 | 235 | CAA54166.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528697 | Chr.11:61690385 - 61919561 on Build GRCh38 | Loss |
|
| nsv343 | Chr.11:61770698 - 61802486 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map